Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101771354-101771613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF8
Entrez Gene ID
2253
Gene Name
fibroblast growth factor 8
Gene Aliases
AIGF, FGF-8, HBGF-8, HH6, KAL6
UniGene
Hs.57710
Gene Chromosome Location
Chr.10: 101770130-101780369 on Build GRCh38
Transcripts
NM_001206389.1 NM_006119.4 NM_033163.3 NM_033164.3 NM_033165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2058603 c.380G>A p.R127Q Chr.10: 101771527-101771527 on Build GRCh38
3699836 c.346A>G p.I116V Chr.10: 101771561-101771561 on Build GRCh38
1317496 c.413G>A p.C138Y Chr.10: 101771494-101771494 on Build GRCh38
4616985 c.410T>C p.I137T Chr.10: 101771497-101771497 on Build GRCh38
7190871 c.356C>T p.T119M Chr.10: 101771551-101771551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112137472 Chr.10: 101771562-101771562 on Build GRCh38
rs773729366 Chr.10: 101771572-101771572 on Build GRCh38
rs777438634 Chr.10: 101771432-101771432 on Build GRCh38
rs774574442 Chr.10: 101771443-101771443 on Build GRCh38
rs754600595 Chr.10: 101771418-101771418 on Build GRCh38
rs774503381 Chr.10: 101771558-101771558 on Build GRCh38
rs767919104 Chr.10: 101771469-101771469 on Build GRCh38
rs769756528 Chr.10: 101771521-101771521 on Build GRCh38
rs904069647 Chr.10: 101771468-101771468 on Build GRCh38
rs746673772 Chr.10: 101771506-101771506 on Build GRCh38
rs367853216 Chr.10: 101771559-101771559 on Build GRCh38
rs762981622 Chr.10: 101771439-101771439 on Build GRCh38
rs767923498 Chr.10: 101771412-101771412 on Build GRCh38
rs200227278 Chr.10: 101771433-101771433 on Build GRCh38
rs137852663 Chr.10: 101771528-101771528 on Build GRCh38
rs3218235 Chr.10: 101771442-101771442 on Build GRCh38
rs943874999 Chr.10: 101771391-101771391 on Build GRCh38
rs201352161 Chr.10: 101771434-101771434 on Build GRCh38
rs575711617 Chr.10: 101771500-101771500 on Build GRCh38
rs745613908 Chr.10: 101771514-101771514 on Build GRCh38
rs1024986845 Chr.10: 101771523-101771523 on Build GRCh38
rs201793069 Chr.10: 101771484-101771484 on Build GRCh38
rs752284353 Chr.10: 101771492-101771492 on Build GRCh38
rs749565857 Chr.10: 101771581-101771582 on Build GRCh38
rs876661329 Chr.10: 101771551-101771551 on Build GRCh38
rs779901890 Chr.10: 101771527-101771527 on Build GRCh38
rs141298356 Chr.10: 101771431-101771431 on Build GRCh38
rs146599295 Chr.10: 101771417-101771417 on Build GRCh38
rs761078377 Chr.10: 101771583-101771583 on Build GRCh38
rs372418903 Chr.10: 101771591-101771591 on Build GRCh38
rs143649717 Chr.10: 101771435-101771435 on Build GRCh38
rs1048331 Chr.10: 101771529-101771529 on Build GRCh38
rs781019978 Chr.10: 101771513-101771513 on Build GRCh38
rs749382906 Chr.10: 101771531-101771531 on Build GRCh38
rs876661330 Chr.10: 101771522-101771522 on Build GRCh38
rs150019367 Chr.10: 101771508-101771508 on Build GRCh38
rs927143916 Chr.10: 101771389-101771389 on Build GRCh38
rs3218234 Chr.10: 101771444-101771444 on Build GRCh38
rs375321211 Chr.10: 101771505-101771505 on Build GRCh38
rs146812350 Chr.10: 101771547-101771547 on Build GRCh38
rs1039842762 Chr.10: 101771445-101771445 on Build GRCh38
rs61730334 Chr.10: 101771509-101771509 on Build GRCh38
rs889652884 Chr.10: 101771387-101771387 on Build GRCh38
rs751003565 Chr.10: 101771475-101771475 on Build GRCh38
rs187295133 Chr.10: 101771427-101771427 on Build GRCh38

View Full Product Details