Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180238277-180238502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK9
Entrez Gene ID
5601
Gene Name
mitogen-activated protein kinase 9
Gene Aliases
JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK
UniGene
Hs.484371
Gene Chromosome Location
Chr.5: 180233594-180292071 on Build GRCh38
Transcripts
NM_002752.4 NM_139068.2 NM_139069.2 NM_139070.2 XM_005265940.4 XM_006714891.2 XM_017009638.1 XM_017009639.1 XM_017009640.1 XM_017009641.1 XM_017009642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5466410 c.1132+9T>G p.? Chr.5: 180238323-180238323 on Build GRCh38
287454 c.1078G>C p.V360L Chr.5: 180238386-180238386 on Build GRCh38
4459398 c.1123C>T p.Q375* Chr.5: 180238341-180238341 on Build GRCh38
1066756 c.1086T>C p.D362D Chr.5: 180238378-180238378 on Build GRCh38
1066757 c.1086T>C p.D362D Chr.5: 180238378-180238378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757234319 Chr.5: 180238441-180238442 on Build GRCh38
rs112353349 Chr.5: 180238469-180238469 on Build GRCh38
rs180730404 Chr.5: 180238411-180238411 on Build GRCh38
rs932089621 Chr.5: 180238384-180238384 on Build GRCh38
rs763418293 Chr.5: 180238403-180238403 on Build GRCh38
rs565556616 Chr.5: 180238430-180238430 on Build GRCh38
rs766792877 Chr.5: 180238405-180238405 on Build GRCh38
rs756137226 Chr.5: 180238428-180238428 on Build GRCh38
rs879278946 Chr.5: 180238359-180238359 on Build GRCh38
rs138473736 Chr.5: 180238354-180238354 on Build GRCh38
rs753506167 Chr.5: 180238383-180238383 on Build GRCh38
rs1037641576 Chr.5: 180238475-180238475 on Build GRCh38
rs777592693 Chr.5: 180238342-180238342 on Build GRCh38
rs55736180 Chr.5: 180238367-180238367 on Build GRCh38
rs114874915 Chr.5: 180238371-180238371 on Build GRCh38
rs769974620 Chr.5: 180238336-180238336 on Build GRCh38
rs367903137 Chr.5: 180238406-180238406 on Build GRCh38
rs759946732 Chr.5: 180238460-180238460 on Build GRCh38
rs774273689 Chr.5: 180238362-180238362 on Build GRCh38
rs770672325 Chr.5: 180238355-180238355 on Build GRCh38
rs911084057 Chr.5: 180238437-180238437 on Build GRCh38
rs770721660 Chr.5: 180238454-180238454 on Build GRCh38
rs751670885 Chr.5: 180238423-180238424 on Build GRCh38
rs781171387 Chr.5: 180238412-180238412 on Build GRCh38
rs781319821 Chr.5: 180238332-180238332 on Build GRCh38
rs373660907 Chr.5: 180238448-180238448 on Build GRCh38
rs759456800 Chr.5: 180238363-180238363 on Build GRCh38
rs531129203 Chr.5: 180238364-180238364 on Build GRCh38
rs371240443 Chr.5: 180238329-180238329 on Build GRCh38
rs923443094 Chr.5: 180238374-180238374 on Build GRCh38
rs982651611 Chr.5: 180238316-180238316 on Build GRCh38
rs35207826 Chr.5: 180238416-180238416 on Build GRCh38
rs534677910 Chr.5: 180238479-180238479 on Build GRCh38

View Full Product Details