Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8056213-8056438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL25
Entrez Gene ID
6370
Gene Name
C-C motif chemokine ligand 25
Gene Aliases
Ckb15, SCYA25, TECK
UniGene
Hs.310511
Gene Chromosome Location
Chr.19: 8052318-8062663 on Build GRCh38
Transcripts
NM_001201359.1 NM_005624.3 XM_011528177.2 XM_017027119.1 XM_017027120.1 XM_017027121.1 XM_017027122.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5883214 c.244G>C p.E82Q Chr.19: 8056418-8056418 on Build GRCh38
1200135 c.192-1G>A p.? Chr.19: 8056365-8056365 on Build GRCh38
3893928 c.254G>A p.R85K Chr.19: 8056428-8056428 on Build GRCh38
6153371 c.156G>A p.Q52Q Chr.19: 8056234-8056234 on Build GRCh38
5057099 c.149G>A p.R50Q Chr.19: 8056227-8056227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763333896 Chr.19: 8056293-8056293 on Build GRCh38
rs760632446 Chr.19: 8056338-8056338 on Build GRCh38
rs762277842 Chr.19: 8056291-8056291 on Build GRCh38
rs77625270 Chr.19: 8056305-8056305 on Build GRCh38
rs764883215 Chr.19: 8056305-8056306 on Build GRCh38
rs772868826 Chr.19: 8056244-8056244 on Build GRCh38
rs759837720 Chr.19: 8056375-8056375 on Build GRCh38
rs776194160 Chr.19: 8056298-8056299 on Build GRCh38
rs373981042 Chr.19: 8056238-8056238 on Build GRCh38
rs747649520 Chr.19: 8056240-8056240 on Build GRCh38
rs948232473 Chr.19: 8056241-8056241 on Build GRCh38
rs750687802 Chr.19: 8056292-8056292 on Build GRCh38
rs1018851890 Chr.19: 8056383-8056383 on Build GRCh38
rs568790594 Chr.19: 8056254-8056254 on Build GRCh38
rs775313102 Chr.19: 8056332-8056332 on Build GRCh38
rs370116383 Chr.19: 8056402-8056402 on Build GRCh38
rs369217231 Chr.19: 8056234-8056234 on Build GRCh38
rs748877843 Chr.19: 8056246-8056246 on Build GRCh38
rs369878493 Chr.19: 8056330-8056330 on Build GRCh38
rs374595746 Chr.19: 8056331-8056331 on Build GRCh38
rs767860072 Chr.19: 8056394-8056394 on Build GRCh38
rs1015020851 Chr.19: 8056290-8056290 on Build GRCh38
rs752280144 Chr.19: 8056345-8056346 on Build GRCh38
rs778243829 Chr.19: 8056416-8056416 on Build GRCh38
rs965011904 Chr.19: 8056396-8056396 on Build GRCh38
rs763101242 Chr.19: 8056371-8056371 on Build GRCh38
rs761501099 Chr.19: 8056385-8056385 on Build GRCh38
rs777202720 Chr.19: 8056307-8056307 on Build GRCh38
rs765359820 Chr.19: 8056283-8056283 on Build GRCh38
rs570205502 Chr.19: 8056290-8056291 on Build GRCh38
rs891467863 Chr.19: 8056313-8056313 on Build GRCh38
rs1009016474 Chr.19: 8056350-8056350 on Build GRCh38
rs759364884 Chr.19: 8056267-8056267 on Build GRCh38
rs964620658 Chr.19: 8056242-8056242 on Build GRCh38
rs751762620 Chr.19: 8056373-8056373 on Build GRCh38
rs748796165 Chr.19: 8056321-8056321 on Build GRCh38
rs781578679 Chr.19: 8056298-8056298 on Build GRCh38
rs766608461 Chr.19: 8056297-8056297 on Build GRCh38
rs376926429 Chr.19: 8056400-8056400 on Build GRCh38
rs769619657 Chr.19: 8056364-8056379 on Build GRCh38
rs200621482 Chr.19: 8056282-8056282 on Build GRCh38
rs201319914 Chr.19: 8056281-8056281 on Build GRCh38
rs762839284 Chr.19: 8056346-8056346 on Build GRCh38
rs774085160 Chr.19: 8056266-8056266 on Build GRCh38
rs769251313 Chr.19: 8056243-8056243 on Build GRCh38
rs773483705 Chr.19: 8056361-8056361 on Build GRCh38
rs768709245 Chr.19: 8056347-8056347 on Build GRCh38

View Full Product Details