Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41221059-41221330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
UniGene
Hs.590970
Gene Chromosome Location
Chr.19: 41219199-41261767 on Build GRCh38
Transcripts
NM_001699.5 NM_021913.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
474777 c.354G>C p.L118F Chr.19: 41221191-41221191 on Build GRCh38
4979035 c.361C>T p.L121L Chr.19: 41221198-41221198 on Build GRCh38
6966165 c.337G>A p.G113R Chr.19: 41221174-41221174 on Build GRCh38
4979034 c.361C>T p.L121L Chr.19: 41221198-41221198 on Build GRCh38
7306391 c.320T>A p.L107Q Chr.19: 41221157-41221157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752898809 Chr.19: 41221234-41221234 on Build GRCh38
rs2271546 Chr.19: 41221292-41221292 on Build GRCh38
rs750304890 Chr.19: 41221169-41221169 on Build GRCh38
rs765391793 Chr.19: 41221207-41221207 on Build GRCh38
rs781434711 Chr.19: 41221186-41221186 on Build GRCh38
rs199544279 Chr.19: 41221283-41221283 on Build GRCh38
rs146822465 Chr.19: 41221131-41221131 on Build GRCh38
rs777785212 Chr.19: 41221170-41221170 on Build GRCh38
rs898531902 Chr.19: 41221130-41221130 on Build GRCh38
rs913110022 Chr.19: 41221142-41221142 on Build GRCh38
rs764751950 Chr.19: 41221167-41221167 on Build GRCh38
rs528480367 Chr.19: 41221162-41221162 on Build GRCh38
rs55682303 Chr.19: 41221221-41221221 on Build GRCh38
rs969922085 Chr.19: 41221214-41221214 on Build GRCh38
rs751357344 Chr.19: 41221173-41221173 on Build GRCh38
rs1006979687 Chr.19: 41221100-41221100 on Build GRCh38
rs927126175 Chr.19: 41221194-41221194 on Build GRCh38
rs927023299 Chr.19: 41221232-41221232 on Build GRCh38
rs992379828 Chr.19: 41221180-41221180 on Build GRCh38
rs749443709 Chr.19: 41221242-41221242 on Build GRCh38
rs771548546 Chr.19: 41221266-41221266 on Build GRCh38
rs866511270 Chr.19: 41221229-41221229 on Build GRCh38
rs760150484 Chr.19: 41221118-41221119 on Build GRCh38
rs890921829 Chr.19: 41221082-41221082 on Build GRCh38
rs1025922950 Chr.19: 41221158-41221158 on Build GRCh38
rs774770336 Chr.19: 41221133-41221133 on Build GRCh38
rs754769605 Chr.19: 41221177-41221177 on Build GRCh38
rs753476920 Chr.19: 41221152-41221152 on Build GRCh38
rs979916017 Chr.19: 41221227-41221227 on Build GRCh38
rs945976266 Chr.19: 41221294-41221294 on Build GRCh38
rs764662403 Chr.19: 41221296-41221296 on Build GRCh38
rs370114405 Chr.19: 41221106-41221106 on Build GRCh38
rs1032706009 Chr.19: 41221083-41221083 on Build GRCh38
rs774773267 Chr.19: 41221269-41221269 on Build GRCh38
rs371995925 Chr.19: 41221275-41221275 on Build GRCh38
rs772705160 Chr.19: 41221298-41221298 on Build GRCh38
rs748402529 Chr.19: 41221201-41221201 on Build GRCh38
rs949895144 Chr.19: 41221168-41221168 on Build GRCh38
rs116592350 Chr.19: 41221139-41221139 on Build GRCh38
rs540192002 Chr.19: 41221215-41221215 on Build GRCh38
rs761470654 Chr.19: 41221293-41221293 on Build GRCh38
rs746215932 Chr.19: 41221270-41221270 on Build GRCh38
rs35202236 Chr.19: 41221172-41221172 on Build GRCh38

View Full Product Details