Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18386335-18386560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF15
Entrez Gene ID
9518
Gene Name
growth differentiation factor 15
Gene Aliases
GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB
UniGene
Hs.616962
Gene Chromosome Location
Chr.19: 18386158-18389176 on Build GRCh38
Transcripts
NM_004864.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1391675 c.269C>T p.T90M Chr.19: 18386458-18386458 on Build GRCh38
5628072 c.240C>T p.L80L Chr.19: 18386429-18386429 on Build GRCh38
4531921 c.183G>A p.E61E Chr.19: 18386372-18386372 on Build GRCh38
4900193 c.267C>T p.L89L Chr.19: 18386456-18386456 on Build GRCh38
4529753 c.163G>A p.E55K Chr.19: 18386352-18386352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562981553 Chr.19: 18386499-18386499 on Build GRCh38
rs751857872 Chr.19: 18386436-18386436 on Build GRCh38
rs752145500 Chr.19: 18386416-18386416 on Build GRCh38
rs762218742 Chr.19: 18386487-18386487 on Build GRCh38
rs560248639 Chr.19: 18386392-18386392 on Build GRCh38
rs1034279886 Chr.19: 18386370-18386370 on Build GRCh38
rs1033182345 Chr.19: 18386498-18386498 on Build GRCh38
rs114827523 Chr.19: 18386458-18386458 on Build GRCh38
rs778245434 Chr.19: 18386358-18386358 on Build GRCh38
rs752235375 Chr.19: 18386534-18386534 on Build GRCh38
rs372819877 Chr.19: 18386427-18386427 on Build GRCh38
rs753738649 Chr.19: 18386424-18386424 on Build GRCh38
rs199975964 Chr.19: 18386495-18386496 on Build GRCh38
rs764881876 Chr.19: 18386501-18386501 on Build GRCh38
rs769807576 Chr.19: 18386384-18386384 on Build GRCh38
rs1031367149 Chr.19: 18386421-18386421 on Build GRCh38
rs745931308 Chr.19: 18386376-18386376 on Build GRCh38
rs772858793 Chr.19: 18386409-18386409 on Build GRCh38
rs757849452 Chr.19: 18386448-18386448 on Build GRCh38
rs780075693 Chr.19: 18386468-18386468 on Build GRCh38
rs776459489 Chr.19: 18386500-18386500 on Build GRCh38
rs771321013 Chr.19: 18386373-18386373 on Build GRCh38
rs775710896 Chr.19: 18386391-18386391 on Build GRCh38
rs768850435 Chr.19: 18386478-18386478 on Build GRCh38
rs747593548 Chr.19: 18386359-18386359 on Build GRCh38
rs77305894 Chr.19: 18386494-18386494 on Build GRCh38
rs770376692 Chr.19: 18386453-18386456 on Build GRCh38
rs1022747118 Chr.19: 18386457-18386457 on Build GRCh38
rs771892633 Chr.19: 18386400-18386400 on Build GRCh38
rs760720564 Chr.19: 18386417-18386417 on Build GRCh38
rs752431818 Chr.19: 18386433-18386433 on Build GRCh38
rs967518704 Chr.19: 18386489-18386489 on Build GRCh38
rs529308097 Chr.19: 18386435-18386435 on Build GRCh38
rs370093085 Chr.19: 18386397-18386397 on Build GRCh38
rs912186813 Chr.19: 18386524-18386524 on Build GRCh38
rs769891685 Chr.19: 18386464-18386464 on Build GRCh38
rs764794071 Chr.19: 18386432-18386432 on Build GRCh38
rs758095463 Chr.19: 18386434-18386434 on Build GRCh38
rs746494128 Chr.19: 18386416-18386422 on Build GRCh38
rs766218944 Chr.19: 18386423-18386423 on Build GRCh38
rs200179968 Chr.19: 18386471-18386471 on Build GRCh38
rs746324703 Chr.19: 18386459-18386459 on Build GRCh38
rs762753931 Chr.19: 18386537-18386537 on Build GRCh38
rs201533332 Chr.19: 18386374-18386374 on Build GRCh38

View Full Product Details