Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111968060-111968333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MERTK
Entrez Gene ID
10461
Gene Name
MER proto-oncogene, tyrosine kinase
Gene Aliases
MER, RP38, Tyro12, c-Eyk, c-mer
UniGene
Hs.306178
Gene Chromosome Location
Chr.2: 111898479-112039946 on Build GRCh38
Transcripts
NM_006343.2 XM_005263565.4 XM_005263568.4 XM_011510490.2 XM_017003164.1 XM_017003165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4701774 c.960G>A p.Q320Q Chr.2: 111968252-111968252 on Build GRCh38
6283105 c.894C>T p.H298H Chr.2: 111968186-111968186 on Build GRCh38
6284487 c.885C>T p.S295S Chr.2: 111968177-111968177 on Build GRCh38
1398795 c.878G>A p.R293H Chr.2: 111968170-111968170 on Build GRCh38
5964771 c.938C>T p.P313L Chr.2: 111968230-111968230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869171331 Chr.2: 111968116-111968117 on Build GRCh38
rs751401651 Chr.2: 111968159-111968159 on Build GRCh38
rs760542995 Chr.2: 111968302-111968302 on Build GRCh38
rs869047507 Chr.2: 111968108-111968109 on Build GRCh38
rs777888162 Chr.2: 111968172-111968172 on Build GRCh38
rs376656831 Chr.2: 111968277-111968277 on Build GRCh38
rs754767582 Chr.2: 111968122-111968122 on Build GRCh38
rs200798420 Chr.2: 111968119-111968119 on Build GRCh38
rs762749580 Chr.2: 111968156-111968156 on Build GRCh38
rs869062260 Chr.2: 111968119-111968120 on Build GRCh38
rs1015124151 Chr.2: 111968190-111968190 on Build GRCh38
rs71226785 Chr.2: 111968122-111968123 on Build GRCh38
rs113538397 Chr.2: 111968107-111968108 on Build GRCh38
rs869028709 Chr.2: 111968118-111968119 on Build GRCh38
rs747858092 Chr.2: 111968170-111968171 on Build GRCh38
rs746294842 Chr.2: 111968131-111968131 on Build GRCh38
rs139569463 Chr.2: 111968097-111968097 on Build GRCh38
rs770737023 Chr.2: 111968266-111968266 on Build GRCh38
rs773927717 Chr.2: 111968274-111968274 on Build GRCh38
rs145602430 Chr.2: 111968148-111968148 on Build GRCh38
rs766215580 Chr.2: 111968157-111968157 on Build GRCh38
rs779211043 Chr.2: 111968204-111968204 on Build GRCh38
rs75918730 Chr.2: 111968107-111968107 on Build GRCh38
rs759345122 Chr.2: 111968162-111968162 on Build GRCh38
rs770131096 Chr.2: 111968211-111968211 on Build GRCh38
rs772859324 Chr.2: 111968244-111968244 on Build GRCh38
rs567714277 Chr.2: 111968288-111968288 on Build GRCh38
rs760731432 Chr.2: 111968108-111968109 on Build GRCh38
rs373446409 Chr.2: 111968292-111968292 on Build GRCh38
rs757475529 Chr.2: 111968201-111968201 on Build GRCh38
rs746561089 Chr.2: 111968175-111968175 on Build GRCh38
rs869029896 Chr.2: 111968117-111968118 on Build GRCh38
rs761363381 Chr.2: 111968147-111968147 on Build GRCh38
rs931976098 Chr.2: 111968144-111968144 on Build GRCh38
rs752968762 Chr.2: 111968101-111968102 on Build GRCh38
rs780578672 Chr.2: 111968230-111968230 on Build GRCh38
rs751328961 Chr.2: 111968115-111968116 on Build GRCh38
rs201359547 Chr.2: 111968087-111968087 on Build GRCh38
rs34072093 Chr.2: 111968170-111968170 on Build GRCh38
rs775953211 Chr.2: 111968138-111968138 on Build GRCh38
rs759853912 Chr.2: 111968097-111968098 on Build GRCh38
rs369655379 Chr.2: 111968169-111968169 on Build GRCh38
rs373198570 Chr.2: 111968261-111968261 on Build GRCh38
rs900547007 Chr.2: 111968143-111968143 on Build GRCh38
rs772663707 Chr.2: 111968137-111968137 on Build GRCh38
rs772369621 Chr.2: 111968212-111968212 on Build GRCh38
rs754002130 Chr.2: 111968179-111968179 on Build GRCh38
rs767470815 Chr.2: 111968166-111968166 on Build GRCh38
rs767285626 Chr.2: 111968286-111968286 on Build GRCh38
rs769412036 Chr.2: 111968235-111968235 on Build GRCh38
rs774712729 Chr.2: 111968124-111968124 on Build GRCh38
rs747537140 Chr.2: 111968231-111968231 on Build GRCh38
rs878853355 Chr.2: 111968225-111968227 on Build GRCh38
rs749583287 Chr.2: 111968109-111968109 on Build GRCh38
rs79025022 Chr.2: 111968101-111968101 on Build GRCh38
rs886871857 Chr.2: 111968236-111968236 on Build GRCh38
rs869157737 Chr.2: 111968123-111968124 on Build GRCh38

View Full Product Details