Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154449860-154450085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL6R
Entrez Gene ID
3570
Gene Name
interleukin 6 receptor
Gene Aliases
CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
UniGene
Hs.135087
Gene Chromosome Location
Chr.1: 154405193-154469450 on Build GRCh38
Transcripts
NM_000565.3 NM_181359.2 XM_005245139.1 XM_005245140.2 XM_006711298.1 XM_006711299.3 XM_017001199.1 XM_017001200.1 XM_017001201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4656112 c.1017C>T p.D339D Chr.1: 154449931-154449931 on Build GRCh38
6058392 c.1006A>T p.T336S Chr.1: 154449920-154449920 on Build GRCh38
3474683 c.1062C>T p.L354L Chr.1: 154449976-154449976 on Build GRCh38
5181899 c.997-10T>C p.? Chr.1: 154449901-154449901 on Build GRCh38
896608 c.1030C>A p.L344I Chr.1: 154449944-154449944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778886483 Chr.1: 154449924-154449924 on Build GRCh38
rs768173607 Chr.1: 154449960-154449960 on Build GRCh38
rs540244501 Chr.1: 154449963-154449963 on Build GRCh38
rs77572132 Chr.1: 154450014-154450014 on Build GRCh38
rs759784340 Chr.1: 154450011-154450011 on Build GRCh38
rs577517640 Chr.1: 154450015-154450015 on Build GRCh38
rs745706538 Chr.1: 154449931-154449931 on Build GRCh38
rs773758849 Chr.1: 154449976-154449976 on Build GRCh38
rs377176989 Chr.1: 154450029-154450029 on Build GRCh38
rs572391772 Chr.1: 154450013-154450013 on Build GRCh38
rs779577954 Chr.1: 154449937-154449937 on Build GRCh38
rs765152700 Chr.1: 154450018-154450018 on Build GRCh38
rs746610451 Chr.1: 154449956-154449956 on Build GRCh38
rs747599074 Chr.1: 154449932-154449932 on Build GRCh38
rs766704226 Chr.1: 154450006-154450006 on Build GRCh38
rs143861748 Chr.1: 154450038-154450038 on Build GRCh38
rs750307810 Chr.1: 154450022-154450022 on Build GRCh38
rs889466996 Chr.1: 154449978-154449978 on Build GRCh38
rs757222842 Chr.1: 154449892-154449892 on Build GRCh38
rs373575595 Chr.1: 154449966-154449966 on Build GRCh38
rs902008358 Chr.1: 154449943-154449943 on Build GRCh38
rs556575220 Chr.1: 154450046-154450046 on Build GRCh38
rs774449360 Chr.1: 154450010-154450010 on Build GRCh38
rs763297632 Chr.1: 154450003-154450003 on Build GRCh38
rs770337126 Chr.1: 154449967-154449967 on Build GRCh38

View Full Product Details