Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101775029-101775254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF8
Entrez Gene ID
2253
Gene Name
fibroblast growth factor 8
Gene Aliases
AIGF, FGF-8, HBGF-8, HH6, KAL6
UniGene
Hs.57710
Gene Chromosome Location
Chr.10: 101770130-101780369 on Build GRCh38
Transcripts
NM_001206389.1 NM_006119.4 NM_033163.3 NM_033164.3 NM_033165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4989458 c.121C>T p.R41W Chr.10: 101775165-101775165 on Build GRCh38
914060 c.92C>T p.A31V Chr.10: 101775194-101775194 on Build GRCh38
1561004 c.106C>A p.L36I Chr.10: 101775180-101775180 on Build GRCh38
7337381 c.76C>T p.P26S Chr.10: 101775210-101775210 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs3218228 Chr.10: 101775088-101775088 on Build GRCh38
rs577352323 Chr.10: 101775120-101775120 on Build GRCh38
rs776130344 Chr.10: 101775164-101775164 on Build GRCh38
rs1032350253 Chr.10: 101775127-101775127 on Build GRCh38
rs762175290 Chr.10: 101775182-101775183 on Build GRCh38
rs746259048 Chr.10: 101775160-101775160 on Build GRCh38
rs1023502394 Chr.10: 101775223-101775223 on Build GRCh38
rs199858724 Chr.10: 101775202-101775202 on Build GRCh38
rs538012694 Chr.10: 101775073-101775073 on Build GRCh38
rs1012456980 Chr.10: 101775214-101775214 on Build GRCh38
rs923038266 Chr.10: 101775048-101775048 on Build GRCh38
rs758099689 Chr.10: 101775130-101775130 on Build GRCh38
rs544494562 Chr.10: 101775194-101775194 on Build GRCh38
rs932128933 Chr.10: 101775094-101775094 on Build GRCh38
rs757584842 Chr.10: 101775124-101775124 on Build GRCh38
rs928359114 Chr.10: 101775232-101775232 on Build GRCh38
rs770042960 Chr.10: 101775161-101775161 on Build GRCh38
rs776087822 Chr.10: 101775208-101775208 on Build GRCh38
rs564054337 Chr.10: 101775072-101775072 on Build GRCh38
rs952053282 Chr.10: 101775228-101775228 on Build GRCh38
rs758593782 Chr.10: 101775104-101775104 on Build GRCh38
rs369975105 Chr.10: 101775053-101775053 on Build GRCh38
rs752575411 Chr.10: 101775093-101775093 on Build GRCh38
rs749761471 Chr.10: 101775177-101775177 on Build GRCh38
rs772729821 Chr.10: 101775193-101775193 on Build GRCh38
rs771618872 Chr.10: 101775188-101775188 on Build GRCh38
rs777982534 Chr.10: 101775114-101775114 on Build GRCh38
rs781205876 Chr.10: 101775156-101775156 on Build GRCh38
rs137852661 Chr.10: 101775168-101775168 on Build GRCh38

View Full Product Details