Gene Transcripts

Gene Symbol
Mta3
Entrez Gene ID
116871
Gene Name
metastasis associated 3
Gene Aliases
1110002J22Rik, mKIAA1266
Chromosome Location
Chr. 17: 83706163 - 83823715 on Build GRCm38
UniGene ID
Mm.277668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171052.1 NP_001164523.1 15 1684
NM_054082.2 NP_473423.1 15 1681
GenBank mRNA AF288138.1 - 15 1635
AK146721.1 - 15 1637
AK146905.1 - 15 1637
AK158866.1 - 14 1547

View Full Product Details