Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253904.1 | NP_001240833.1 | 21 | 2773 | |
NM_001253905.1 | NP_001240834.1 | 17 | 2327 | ||
NM_011424.3 | NP_035554.3 | 21 | 2938 | ||
GenBank mRNA | AF113002.1 | - | 17 | 2319 | |
AF125671.1 | - | 21 | 2773 | ||
AK147394.1 | - | 21 | 2739 | ||
AK158191.1 | - | 4 | 864 | ||
AK170097.1 | - | 5 | 1370 |