Gene Transcripts

Gene Symbol
Nrros
Entrez Gene ID
224109
Gene Name
negative regulator of reactive oxygen species
Gene Aliases
E430025L02Rik, Lrcc33, Lrrc33
Chromosome Location
Chr. 16: 32142785 - 32165589 on Build GRCm38
UniGene ID
Mm.33498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146069.4 NP_666181.2 4 3098
GenBank mRNA AK028367.1 - 2 2050
AK042502.1 - 2 1900
AK084858.1 - 4 3101
AK088770.1 - 2 1819
AK128897.1 - 3 1938
AK167934.1 - 3 1968
BC027411.1 - 3 1948 4192329

View Full Product Details