Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081071.2 | NP_001074540.1 | 3 | 385 | |
NM_001177967.1 | NP_001171438.1 | 3 | 364 | ||
NM_001177968.1 | NP_001171439.1 | 3 | 353 | ||
GenBank mRNA | AK144580.1 | - | 3 | 328 | |
BC147499.1 | - | 3 | 303 | 9056361 | |
BC150884.1 | - | 3 | 292 | 9087795 | |
BC158076.1 | - | 3 | 324 | 9088168 | |
BC158079.1 | - | 3 | 324 | 9088171 |