Gene Transcripts
- Gene Symbol
- Lrsam1
- Entrez Gene ID
- 227738
- Gene Name
- leucine rich repeat and sterile alpha motif containing 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 32925215 - 32961654 on Build GRCm38
- UniGene ID
- Mm.248037
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_199302.2 |
NP_955006.1
|
16, 17 |
1494 |
|
GenBank mRNA |
BC049146.1 |
- |
16, 17 |
1490 |
|
View Full Product Details