Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033409.3 | NP_001028581.1 | 16 | 1593 | |
GenBank mRNA | AK052873.1 | - | 11 | 793 | |
AK085901.1 | - | 16 | 1552 | ||
AK137380.1 | - | 16 | 1594 | ||
BC141210.1 | - | 16 | 1545 | 9055822 |