Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025570.2 | NP_079846.1 | 4 | 380 | |
GenBank mRNA | AB049645.1 | - | 4 | 353 | |
AK004053.1 | - | 4 | 365 | ||
AK011341.1 | - | 4 | 381 | ||
AK015199.1 | - | 364 | |||
AK168665.1 | - | 4 | 354 | ||
BC039985.1 | - | 4 | 350 | 1244957 |