Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025649.3 | NP_079925.2 | 3 | 647 | |
GenBank mRNA | AK002377.1 | - | 3 | 633 | |
AK004224.1 | - | 3 | 601 | ||
AK007974.1 | - | 3 | 599 | ||
AK156569.1 | - | 3 | 650 | ||
AK220322.1 | - | 3 | 616 | ||
BC005768.1 | - | 3 | 605 | 2649017 |