Gene Transcripts

Gene Symbol
Mphosph10
Entrez Gene ID
67973
Gene Name
M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein)
Gene Aliases
2810453H10Rik, 5730405D16Rik, AI326008
Chromosome Location
Chr. 7: 64376541 - 64392236 on Build GRCm38
UniGene ID
Mm.26973
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026483.2 NP_080759.2 8 1546
GenBank mRNA AK008785.1 - 5 476
AK013337.1 - 8 1534
BC018177.1 - 8 1540

View Full Product Details