Gene Transcripts

Gene Symbol
Morc4
Entrez Gene ID
75746
Gene Name
microrchidia 4
Gene Aliases
1600017G11Rik, 5630401M14Rik, AA117449, Zcwcc2
Chromosome Location
Chr. X: 139821635 - 139871692 on Build GRCm38
UniGene ID
Mm.23269
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193309.1 NP_001180238.1 11 1599
NM_029413.4 NP_083689.2 11 1599
GenBank mRNA AK017472.1 - 11 1568
AK032807.1 - 11 1600
BC036164.1 - 5 486
BC039283.1 - 4 311
BC098483.1 - 11 1554 30022418
BC132153.1 - 11 1453 40130430
BC132497.1 - 11 1453 40130774

View Full Product Details