Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256100.1 | NP_001243029.1 | 4 | 766 | |
NM_001256101.1 | NP_001243030.1 | 4 | 755 | ||
NM_001256102.1 | NP_001243031.1 | 3 | 595 | ||
NM_029581.4 | NP_083857.1 | 3 | 606 | ||
GenBank mRNA | AK012733.1 | - | 4 | 733 | |
AK147090.1 | - | 3 | 584 | ||
BC052439.1 | - | 3 | 560 | 6406303 | |
BC115684.1 | - | 3 | 547 | 40104230 | |
BC115685.1 | - | 4 | 718 | 40104232 |