Gene Transcripts

Gene Symbol
Ctgf
Entrez Gene ID
14219
Gene Name
connective tissue growth factor
Gene Aliases
Ccn2, Fisp12, Hcs24, fisp-12
Chromosome Location
Chr. 10: 24595442 - 24598683 on Build GRCm38
UniGene ID
Mm.390287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010217.2 NP_034347.2 4 860
GenBank mRNA BC006783.1 - 4 781
BQ917383.1 - 4 798
CJ055589.1 - 2 239
M70642.1 - 4 713
M80263.1 - 4 779

View Full Product Details