Gene Transcripts

Gene Symbol
Nfyc
Entrez Gene ID
18046
Gene Name
nuclear transcription factor-Y gamma
Gene Aliases
Chromosome Location
Chr. 4: 120757435 - 120831579 on Build GRCm38
UniGene ID
Mm.435590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048168.2 NP_001041633.1 8 881
NM_001277095.1 NP_001264024.1 7 840
NM_008692.4 NP_032718.2 8 954
GenBank mRNA AB006181.1 - 7 734
AK011327.1 - 8 910
AK156353.1 - 7 694
AK186792.1 - 1 16
AK216147.1 - 1 16
AK219678.1 - 1 16
BC020117.1 - 8 880 4009737
BC053723.1 - 8 884 30053020
BC085261.1 - 8 880 6397760
BM461330.1 - 6 585

View Full Product Details