Gene Transcripts
- Gene Symbol
- Nfyc
- Entrez Gene ID
- 18046
- Gene Name
- nuclear transcription factor-Y gamma
- Gene Aliases
- Chromosome Location
- Chr. 4: 120757435 - 120831579 on Build GRCm38
- UniGene ID
- Mm.435590
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001048168.2 |
NP_001041633.1
|
8, 9 |
963 |
|
NM_001277095.1 |
NP_001264024.1
|
7, 8 |
922 |
|
NM_008692.4 |
NP_032718.2
|
8, 9 |
1036 |
|
GenBank mRNA |
AB006181.1 |
- |
7, 8 |
816 |
|
AK011327.1 |
- |
8, 9 |
992 |
|
AK156353.1 |
- |
7, 8 |
776 |
|
AK186792.1 |
- |
1, 2 |
98 |
|
AK216147.1 |
- |
1, 2 |
98 |
|
AK219678.1 |
- |
1, 2 |
98 |
|
BC020117.1 |
- |
8, 9 |
962 |
4009737 |
BC085261.1 |
- |
8, 9 |
962 |
6397760 |
BM461330.1 |
- |
6, 7 |
667 |
|
U62297.1 |
- |
7, 8 |
816 |
|
View Full Product Details