Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173012.3 | NP_766600.2 | 8 | 1531 | |
GenBank mRNA | AK163507.1 | - | 8 | 1385 | |
BC050979.1 | - | 4 | 436 | ||
BC055685.1 | - | 8 | 1531 | 6403034 | |
BE648035.1 | - | 3 | 318 |