Gene Transcripts

Gene Symbol
Lpar5
Entrez Gene ID
381810
Gene Name
lysophosphatidic acid receptor 5
Gene Aliases
GPR93, Gm1072, Gpr92, LPA5
Chromosome Location
Chr. 6: 125067842 - 125086129 on Build GRCm38
UniGene ID
Mm.333386
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163268.1 NP_001156740.1 2 1225
NM_001163269.1 NP_001156741.1 2 1160
GenBank mRNA BC116702.1 - 2 1125
BC117528.1 - 2 1125 40104864

View Full Product Details