Gene Transcripts

Gene Symbol
Mast3
Entrez Gene ID
546071
Gene Name
microtubule associated serine/threonine kinase 3
Gene Aliases
BC024265, mKIAA0561
Chromosome Location
Chr. 8: 70778117 - 70805068 on Build GRCm38
UniGene ID
Mm.250731
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199308.2 NP_955012.2 26 3862
GenBank mRNA AK140821.1 - 26 3878
AK172971.1 - 6 1074
BC024265.1 - 8 1588

View Full Product Details