Gene Transcripts
- Gene Symbol
- Ndufs5
- Entrez Gene ID
- 595136
- Gene Name
- NADH dehydrogenase (ubiquinone) Fe-S protein 5
- Gene Aliases
- AI256693
- Chromosome Location
- Chr. 4: 123712710 - 123718186 on Build GRCm38
- UniGene ID
- Mm.42805
- Species
- Mus musculus
- Gene Symbol
- BC002163
- Entrez Gene ID
- 170658
- Gene Name
- NADH dehydrogenase Fe-S protein 5 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 16: 42955580 - 42956079 on Build GRCm38
- UniGene ID
- Mm.375824
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC002163.1 |
- |
1 |
285 |
|
View Full Product Details