Gene Transcripts

Gene Symbol
Smco1
Entrez Gene ID
69576
Gene Name
single-pass membrane protein with coiled-coil domains 1
Gene Aliases
2310010M20Rik
Chromosome Location
Chr. 16: 32271609 - 32274779 on Build GRCm38
UniGene ID
Mm.432545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183283.2 NP_899106.2 3 536
GenBank mRNA AK009281.1 - 3 648
BC125555.1 - 2 494 40130070
BC131954.1 - 2 494

View Full Product Details