Gene Transcripts

Gene Symbol
Nos1ap
Entrez Gene ID
70729
Gene Name
nitric oxide synthase 1 (neuronal) adaptor protein
Gene Aliases
6330408P19Rik, Capon
Chromosome Location
Chr. 1: 170317414 - 170589849 on Build GRCm38
UniGene ID
Mm.487078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109985.1 NP_001103455.1 4, 5 808
NM_027528.2 NP_081804.1 4, 5 625
GenBank mRNA AK138471.1 - 4, 5 626
AK162919.1 - 4, 5 626

View Full Product Details