Gene Transcripts

Gene Symbol
Zfp263
Entrez Gene ID
287076
Gene Name
zinc finger protein 263
Gene Aliases
Chromosome Location
Chr. 10: 12023677 - 12035182 on Build Rnor_6.0
UniGene ID
Rn.45317
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105763.1 NP_001099233.1 5 1131
XM_006245830.2 XP_006245892.1 5 5803
XM_006245831.2 XP_006245893.1 3 5547

View Full Product Details