Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286183.1 | NP_001273112.1 | 3 | 646 | |
XM_003750804.2 | XP_003750852.1 | 3 | 774 | ||
XM_003750806.2 | XP_003750854.1 | 2 | 605 | ||
XM_006246153.2 | XP_006246215.1 | 2 | 2944 | ||
XM_006246158.2 | XP_006246220.1 | 3 | 619 | ||
XM_006246159.2 | XP_006246221.1 | 3 | 599 | ||
XM_006246161.1 | XP_006246223.1 | 2 | 533 | ||
XM_008767653.1 | XP_008765875.1 | 3 | 618 | ||
GenBank mRNA | FQ228105.1 | - | 3 | 632 |