Gene Transcripts

Gene Symbol
Lppr5
Entrez Gene ID
310812
Gene Name
lipid phosphate phosphatase-related protein type 5
Gene Aliases
PRG-5, Pap2d, RGD1309567
Chromosome Location
Chr. 2: 219895141 - 220102998 on Build Rnor_6.0
UniGene ID
Rn.225021
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107720.1 NP_001101190.1 3 1207
XM_008761462.1 XP_008759684.1 3 2414
GenBank mRNA EU792473.1 - 3 1207

View Full Product Details