Gene Transcripts

Gene Symbol
Nhs
Entrez Gene ID
317494
Gene Name
Nance-Horan syndrome (congenital cataracts and dental anomalies)
Gene Aliases
RGD1561818
Chromosome Location
Chr. X: 34313566 - 34675912 on Build Rnor_6.0
UniGene ID
Rn.224910
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191733.1 NP_001178662.1 6 4398
XM_008773173.1 XP_008771395.1 7 4862

View Full Product Details