Gene Transcripts

Gene Symbol
Mrpl48
Entrez Gene ID
293149
Gene Name
mitochondrial ribosomal protein L48
Gene Aliases
Chromosome Location
Chr. 1: 165563788 - 165606375 on Build Rnor_6.0
UniGene ID
Rn.107172
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106282.1 NP_001099752.1 7 622
GenBank mRNA FQ212514.1 - 7 819
FQ214311.1 - 7 671

View Full Product Details