Gene Transcripts

Gene Symbol
Olr834
Entrez Gene ID
298396
Gene Name
olfactory receptor 834
Gene Aliases
Chromosome Location
Chr. 5: 59207120 - 59208058 on Build Rnor_6.0
UniGene ID
Rn.143214
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000407.1 NP_001000407.1 1 518

View Full Product Details