Gene Transcripts

Gene Symbol
Olr1474
Entrez Gene ID
405285
Gene Name
olfactory receptor 1474
Gene Aliases
Chromosome Location
Chr. 10: 60125764 - 60126702 on Build Rnor_6.0
UniGene ID
Rn.142095
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000943.1 NP_001000943.1 1 448

View Full Product Details