Gene Transcripts

Gene Symbol
Lrit2
Entrez Gene ID
498594
Gene Name
leucine-rich repeat, immunoglobulin-like and transmembrane domains 2
Gene Aliases
RGD1564668
Chromosome Location
Chr. 16: 14318365 - 14324348 on Build Rnor_6.0
UniGene ID
Rn.228222
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191938.1 NP_001178867.1 3 959
XM_006252787.2 XP_006252849.1 3 2242

View Full Product Details