Gene Transcripts

Gene Symbol
Blnk
Entrez Gene ID
499356
Gene Name
B-cell linker
Gene Aliases
Chromosome Location
Chr. 1: 260187023 - 260254490 on Build Rnor_6.0
UniGene ID
Rn.32684
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025767.1 NP_001020938.1 2 202
GenBank mRNA BC098790.1 - 2 202

View Full Product Details