Gene Transcripts

Gene Symbol
RGD1560608
Entrez Gene ID
500724
Gene Name
similar to novel protein
Gene Aliases
Chromosome Location
Chr. 6: 135856218 - 135861059 on Build Rnor_6.0
UniGene ID
Rn.53684
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109280.1 NP_001102750.1 4 415

View Full Product Details