Gene Transcripts

Gene Symbol
Odf3b
Entrez Gene ID
500917
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
RGD1564322
Chromosome Location
Chr. 7: 130347945 - 130351571 on Build Rnor_6.0
UniGene ID
Rn.76400
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191992.1 NP_001178921.1 1, 2 108
XM_006242238.2 XP_006242300.1 2, 3 751
XM_008765776.1 XP_008763998.1 1, 2 491

View Full Product Details