Gene Transcripts

Gene Symbol
BAD
Entrez Gene ID
572
Gene Name
BCL2 associated agonist of cell death
Gene Aliases
BBC2, BCL2L8
Chromosome Location
Chr.11: 64269828 - 64284704 on Build GRCh38
Uni Gene
Hs.370254
Interrogated Sequence Translated Protein
RefSeq NM_004322.3 NP_004313.1
NM_032989.2 NP_116784.1
GenBank mRNA AB451254.1 -
AB451378.1 -
AF021792.1 -
AF031523.1 -
AK023420.1 -
AK291863.1 -
AK303051.1 -
AK309150.1 -
AW014069.1 -
BC001901.2 -
BC095431.1 -
BE255791.1 -
BG435829.1 -
BG748336.1 -
BT006678.1 -
CR541935.1 -
CR541959.1 -
ENST00000394532.0 -
U66879.1 -

View Full Product Details