Gene Transcripts

Gene Symbol
GDF15
Entrez Gene ID
9518
Gene Name
growth differentiation factor 15
Gene Aliases
GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB
Chromosome Location
Chr.19: 18386158 - 18389176 on Build GRCh38
Uni Gene
Hs.616962
Interrogated Sequence Translated Protein
RefSeq NM_004864.2 NP_004855.2
GenBank mRNA AB000584.1 -
AF003934.1 -
AF019770.1 -
AF173860.1 -
AK225431.1 -
AK291530.1 -
BC000529.2 -
BC008962.2 -
BT019465.1 -
ENST00000252809.0 -
U51731.1 -
U88323.1 -

View Full Product Details