Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001098209.1 | NP_001091679.1 |
NM_001098210.1 | NP_001091680.1 | |
NM_001904.3 | NP_001895.1 | |
XM_005264886.2 | XP_005264943.1 | |
XM_006712983.1 | XP_006713046.1 | |
XM_006712984.1 | XP_006713047.1 | |
XM_006712985.1 | XP_006713048.1 | |
XM_017005738.1 | XP_016861227.1 | |
GenBank mRNA | AB062292.1 | - |
AB451264.1 | - | |
AB451392.1 | - | |
AK095242.1 | - | |
AK289932.1 | - | |
AK294778.1 | - | |
AK296790.1 | - | |
AK299954.1 | - | |
AK314776.1 | - | |
BC058926.1 | - | |
BX648430.1 | - | |
DA216720.1 | - | |
ENST00000349496.0 | - | |
U38440.1 | - | |
X87838.1 | - | |
Z19054.1 | - |