Gene Transcripts

Gene Symbol
SP1
Entrez Gene ID
6667
Gene Name
Sp1 transcription factor
Gene Aliases
-
Chromosome Location
Chr.12: 53380195 - 53416446 on Build GRCh38
Uni Gene
Hs.620754
Interrogated Sequence Translated Protein
RefSeq NM_001251825.1 NP_001238754.1
NM_003109.1 NP_003100.1
NM_138473.2 NP_612482.2
XM_011538696.2 XP_011536998.1
GenBank mRNA AB209678.1 -
AF252284.1 -
AF255682.1 -
AI282745.1 -
AI561005.1 -
AJ272134.1 -
AK311995.1 -
AL442093.1 -
AW028976.1 -
BC012008.1 -
BC043224.1 -
BC062539.1 -
BC078158.1 -
BF434304.1 -
BF436290.1 -
BF671559.1 -
BM463205.1 -
BQ072120.1 -
BQ431320.1 -
BQ774060.1 -
CD104746.1 -
FJ949572.1 -
FN908228.1 -
J03133.1 -

View Full Product Details