Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001014431.1 | NP_001014431.1 |
NM_001014432.1 | NP_001014432.1 | |
NM_005163.2 | NP_005154.2 | |
XM_005267401.1 | XP_005267458.1 | |
XM_017021075.1 | XP_016876564.1 | |
XM_017021076.1 | XP_016876565.1 | |
XM_017021077.1 | XP_016876566.1 | |
XM_017021078.1 | XP_016876567.1 | |
GenBank mRNA | AB451242.1 | - |
AB451367.1 | - | |
AK094287.1 | - | |
AK122894.1 | - | |
AK131465.1 | - | |
AK299310.1 | - | |
AK314256.1 | - | |
BC000479.2 | - | |
BC001737.1 | - | |
BC063408.1 | - | |
BC084538.1 | - | |
BX647722.1 | - | |
BX648205.1 | - | |
CN414251.1 | - | |
ENST00000349310.0 | - | |
KF836747.1 | - | |
KF836748.1 | - | |
KF836749.1 | - | |
KF836750.1 | - | |
KU177898.1 | - | |
KU177899.1 | - | |
M63167.1 | - | |
X61037.1 | - |