Gene Transcripts

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
Chromosome Location
Chr.19: 41219199 - 41261767 on Build GRCh38
Uni Gene
Hs.590970
Interrogated Sequence Translated Protein
RefSeq NM_001278599.1 NP_001265528.1
NM_001699.5 NP_001690.2
NM_021913.4 NP_068713.2
GenBank mRNA AA021223.1 -
AI300772.1 -
AI309979.1 -
AK309139.1 -
AK310881.1 -
AL556721.3 -
BC032229.1 -
CA437238.1 -
DA478638.1 -
ENST00000359092.0 -
HY045208.1 -
M76125.1 -
S65125.1 -
X57019.1 -
X66029.1 -

View Full Product Details