Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001099856.4 | NP_001093326.2 |
NM_001099857.2 | NP_001093327.1 | |
NM_001145255.2 | NP_001138727.1 | |
NM_001321396.1 | NP_001308325.1 | |
NM_001321397.1 | NP_001308326.1 | |
NM_003639.4 | NP_003630.1 | |
GenBank mRNA | AF062089.1 | - |
AF074382.1 | - | |
AF091453.1 | - | |
AF261086.1 | - | |
AK000593.1 | - | |
AK308366.1 | - | |
AY114157.1 | - | |
AY429546.1 | - | |
AY429547.1 | - | |
AY429548.1 | - | |
BC000299.2 | - | |
BC012114.1 | - | |
BC046922.1 | - | |
BC050612.2 | - | |
BM473416.1 | - | |
BT019621.1 | - | |
CA916723.1 | - | |
DC410130.1 | - |