Gene Transcripts

Gene Symbol
FGF8
Entrez Gene ID
2253
Gene Name
fibroblast growth factor 8
Gene Aliases
AIGF, FGF-8, HBGF-8, HH6, KAL6
Chromosome Location
Chr.10: 101770130 - 101780369 on Build GRCh38
Uni Gene
Hs.57710
Interrogated Sequence Translated Protein
RefSeq NM_001206389.1 NP_001193318.1
NM_006119.4 NP_006110.1
NM_033163.3 NP_149353.1
NM_033164.3 NP_149354.1
NM_033165.3 NP_149355.1
GenBank mRNA AB014615.1 -
BC069106.1 -
BC128235.1 -
BC128236.1 -
CN369230.1 -
ENST00000320185.0 -
U36223.1 -
U46211.1 -
U46212.1 -
U46213.1 -
U56978.1 -

View Full Product Details