Gene Transcripts

Gene Symbol
OGFR
Entrez Gene ID
11054
Gene Name
opioid growth factor receptor
Gene Aliases
-
Chromosome Location
Chr.20: 62804825 - 62814000 on Build GRCh38
Uni Gene
Hs.67896
Interrogated Sequence Translated Protein
RefSeq NM_007346.2 NP_031372.2
GenBank mRNA AF109134.1 -
AF172449.1 -
AF172450.1 -
AF172451.1 -
AF172452.1 -
AF172453.1 -
AK022234.1 -
AK024485.1 -
AK314696.1 -
BC008768.2 -
BC014137.2 -
BC032666.1 -
BM551600.1 -
ENST00000290291.0 -

View Full Product Details