Gene Transcripts

Gene Symbol
ANGPT2
Entrez Gene ID
285
Gene Name
angiopoietin 2
Gene Aliases
AGPT2, ANG2
Chromosome Location
Chr.8: 6499651 - 6563420 on Build GRCh38
Uni Gene
Hs.583870
Interrogated Sequence Translated Protein
RefSeq NM_001118887.1 NP_001112359.1
NM_001118888.1 NP_001112360.1
NM_001147.2 NP_001138.1
XM_017013318.1 XP_016868807.1
GenBank mRNA AB009865.1 -
AF004327.1 -
AF187858.1 -
AF218015.1 -
AJ289780.1 -
AJ289781.1 -
AK075219.1 -
AK225698.1 -
AK290070.1 -
AK310171.1 -
AK312541.1 -
BC022490.1 -
BC126200.1 -
BC126202.1 -
BC143902.1 -
DA829327.1 -
ENST00000325203.0 -

View Full Product Details