Gene Transcripts

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
Chromosome Location
Chr.5: 159314783 - 159330473 on Build GRCh38
Uni Gene
Hs.674
Interrogated Sequence Translated Protein
RefSeq NM_002187.2 NP_002178.2
GenBank mRNA AF180563.1 -
AY046592.1 -
AY046593.1 -
BC067498.1 -
BC067499.1 -
BC067500.1 -
BC067501.1 -
BC067502.1 -
BC074723.2 -
EF173865.1 -
ENST00000231228.0 -
M65272.1 -
M65290.1 -

View Full Product Details